N-Methoxy-N-methylformamide 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245265-10GEA 3160 DKK
BLDPBD245265-10G BLDPBD245265-5G BLDPBD245265-1G BLDPBD245265-250MG
N-Methoxy-N-methylformamide 95%
N-Methoxy-N-methylformamide

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₃H₇NO₂
Molekylvægt: 89,09 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD11226240
CAS-nummer: 32117-82-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR