(R)-3-(3,4-Dihydroxyphenyl)-2-hydroxypropanoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245214-100MGEA 791 DKK
BLDPBD245214-100MG
(R)-3-(3,4-Dihydroxyphenyl)-2-hydroxypropanoic acid 98%
(R)-3-(3,4-Dihydroxyphenyl)-2-hydroxypropanoic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀O₅
Molekylvægt: 198,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09038703
CAS-nummer: 76822-21-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR