3-Bromo-4-chlorobenzeneacetic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245114-10GEA 1560 DKK
BLDPBD245114-10G BLDPBD245114-25G BLDPBD245114-5G BLDPBD245114-250MG BLDPBD245114-100G BLDPBD245114-1G
3-Bromo-4-chlorobenzeneacetic acid 97%
3-Bromo-4-chlorobenzeneacetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆BrClO₂
Molekylvægt: 249,489 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18390582
CAS-nummer: 90004-80-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR