3-Ethynylaniline hydrochloride 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245058-5GEA 109 DKK
BLDPBD245058-5G BLDPBD245058-25G BLDPBD245058-100G BLDPBD245058-10G BLDPBD245058-500G BLDPBD245058-1G
3-Ethynylaniline hydrochloride 95%
3-Ethynylaniline hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₈ClN
Molekylvægt: 153,6 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12913913
CAS-nummer: 207226-02-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR