Methyl 2-amino-5-fluoronicotinate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245029-250MGEA 2440 DKK
BLDPBD245029-250MG BLDPBD245029-100MG BLDPBD245029-1G
Methyl 2-amino-5-fluoronicotinate 98%
Methyl 2-amino-5-fluoronicotinate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₇H₇FN₂O₂
Molekylvægt: 170,14 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18261243
CAS-nummer: 1211535-54-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR