4,4-dinitro-2,2-Bipyridine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD245016-1GEA 1050 DKK
BLDPBD245016-1G BLDPBD245016-250MG BLDPBD245016-100MG
4,4-dinitro-2,2-Bipyridine 98%
4,4-dinitro-2,2-Bipyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₀H₆N₄O₄
Molekylvægt: 246,17 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06657644
CAS-nummer: 18511-72-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR