tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD24501-250MGEA 140 DKK
BLDPBD24501-250MG BLDPBD24501-5G BLDPBD24501-1G BLDPBD24501-100G BLDPBD24501-25G BLDPBD24501-100MG BLDPBD24501-10G
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate 97%
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₆H₂₄N₂O₂
Molekylvægt: 276,3740 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05861405
CAS-nummer: 170011-57-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR