3-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244883-250MGEA 931 DKK
BLDPBD244883-250MG BLDPBD244883-1G BLDPBD244883-100MG BLDPBD244883-5G
3-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 95%
3-Ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₂₀BNO₃
Molekylvægt: 249,11 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11878280
CAS-nummer: 1171892-40-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR