(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-phenylpropanoic acid 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244641-50MGEA 1110 DKK
BLDPBD244641-50MG BLDPBD244641-25MG
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-phenylpropanoic acid 95%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-phenylpropanoic acid
Formel: C₂₄H₂₁NO₄
Molekylvægt: 387,43 g/mol
MDL Number: MFCD11052914
CAS-nummer: 1217722-24-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR