H-β-HomoAsp·HCl 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244619-100MGEA 652 DKK
BLDPBD244619-100MG BLDPBD244619-250MG BLDPBD244619-1G
H-β-HomoAsp·HCl 98%
H-β-HomoAsp·HCl

Storage condition: Inert atmosphere, 2-8°C

Formel: C₅H₉NO₄·HCl
Molekylvægt: 183,6 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD01862859
CAS-nummer: 336182-10-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR