1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244492-5GEA 2710 DKK
BLDPBD244492-5G BLDPBD244492-1G
1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene 98%
1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₂₇H₃₆N₂
Molekylvægt: 388,60 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD07781991
CAS-nummer: 244187-81-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR