5-Chloro-1-methyl-4-nitro-1H-pyrazole 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244205-100MGEA 435 DKK
BLDPBD244205-100MG BLDPBD244205-5G BLDPBD244205-1G BLDPBD244205-250MG
5-Chloro-1-methyl-4-nitro-1H-pyrazole 97%
5-Chloro-1-methyl-4-nitro-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₄H₄ClN₃O₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD21607270
CAS-nummer: 42098-25-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR