7-Fluorobenzo[d]thiazol-2-amine 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244124-250MGEA 1190 DKK
BLDPBD244124-250MG BLDPBD244124-100MG BLDPBD244124-1G
7-Fluorobenzo[d]thiazol-2-amine 97%
7-Fluorobenzo[d]thiazol-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₅FN₂S
Molekylvægt: 168,191 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11110958
CAS-nummer: 20358-08-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR