tert-Butyl octahydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244093-50MGEA 1050 DKK
BLDPBD244093-50MG BLDPBD244093-250MG BLDPBD244093-100MG
tert-Butyl octahydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate 96%
tert-Butyl octahydro-1H-pyrrolo[3,2-b]pyridine-1-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₁₂H₂₂N₂O₂
Molekylvægt: 226,31 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18382710
CAS-nummer: 1211586-14-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR