7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD244048-100MGEA 303 DKK
BLDPBD244048-100MG BLDPBD244048-25G BLDPBD244048-5G BLDPBD244048-1G BLDPBD244048-250MG
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline 97%
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₅H₁₈BNO₂
Molekylvægt: 255,12 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD08063115
CAS-nummer: 851985-81-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR