Phenazine-1-carboxamide 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD243981-1GEA 4060 DKK
BLDPBD243981-1G BLDPBD243981-250MG BLDPBD243981-100MG BLDPBD243981-5G
Phenazine-1-carboxamide 98%
Phenazine-1-carboxamide

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₉N₃O
Molekylvægt: 223,23 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01708506
CAS-nummer: 550-89-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR