3-Methyl-3-oxetanmethanol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD2439-25GEA 101 DKK
BLDPBD2439-25G BLDPBD2439-100G BLDPBD2439-5G BLDPBD2439-1000G BLDPBD2439-500G BLDPBD2439-10G
3-Methyl-3-oxetanmethanol 97%
3-Methyl-3-oxetanmethanol

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₁₀O₂
Molekylvægt: 102,13 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00010273
CAS-nummer: 3143-02-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR