2-(Ethoxycarbonyl)furan-5-boronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD243764-5GEA 3690 DKK
BLDPBD243764-5G
2-(Ethoxycarbonyl)furan-5-boronic acid 98%
2-(Ethoxycarbonyl)furan-5-boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₇H₉BO₅
Molekylvægt: 183,9544 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD10696632
CAS-nummer: 1150114-44-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR