4-Bromo-1H-pyrrole-2-carboxamide 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD243447-250MGEA 512 DKK
BLDPBD243447-250MG BLDPBD243447-100MG BLDPBD243447-1G BLDPBD243447-5G
4-Bromo-1H-pyrrole-2-carboxamide 97%
4-Bromo-1H-pyrrole-2-carboxamide

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₅BrN₂O
Molekylvægt: 189,01 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11504939
CAS-nummer: 196106-96-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR