3-Bromo-2-chloro-5-thiophenesulphonyl chloride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD2405-250MGEA 148 DKK
BLDPBD2405-250MG BLDPBD2405-5G
3-Bromo-2-chloro-5-thiophenesulphonyl chloride 97%
3-Bromo-2-chloro-5-thiophenesulphonyl chloride

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₄HBrCl₂O₂S₂
Molekylvægt: 295,9895 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00052094
CAS-nummer: 166964-35-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR