Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD239942-100MGEA 241 DKK
BLDPBD239942-100MG BLDPBD239942-1G BLDPBD239942-250MG
Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate 98%
Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₈H₂₁BO₄
Molekylvægt: 312,168 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD18783177
CAS-nummer: 736989-93-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR