Methyl-2-fluoro-5-methylbenzoate 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD236696-100GEA 3310 DKK
BLDPBD236696-100G BLDPBD236696-10G BLDPBD236696-1G BLDPBD236696-5G BLDPBD236696-25G
Methyl-2-fluoro-5-methylbenzoate 98%
Methyl-2-fluoro-5-methylbenzoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉FO₂
Molekylvægt: 168,1700 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06203924
CAS-nummer: 2967-93-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR