2,4,6-Triisopropylphenylboronic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD236236-5GEA 225 DKK
BLDPBD236236-5G BLDPBD236236-250MG BLDPBD236236-25G BLDPBD236236-100G BLDPBD236236-1G BLDPBD236236-10G
2,4,6-Triisopropylphenylboronic acid 98%
2,4,6-Triisopropylphenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₅H₂₅BO₂
Molekylvægt: 248,16 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD02683099
CAS-nummer: 154549-38-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR