6-Hydroxyisoindolin-1-one 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD235521-100MGEA 357 DKK
BLDPBD235521-100MG BLDPBD235521-1G BLDPBD235521-250MG BLDPBD235521-5G
6-Hydroxyisoindolin-1-one 95%
6-Hydroxyisoindolin-1-one

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₈H₇NO₂
Molekylvægt: 149,147 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08276013
CAS-nummer: 659737-57-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR