2-Ethyl-5-methylthiophene 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD235329-250MGEA 46.6 DKK
BLDPBD235329-250MG BLDPBD235329-5G BLDPBD235329-1G
2-Ethyl-5-methylthiophene 98%
2-Ethyl-5-methylthiophene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₁₀S
Molekylvægt: 126,22 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00053128
CAS-nummer: 40323-88-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR