2-(Propan-2-yl)morpholine 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD233814-1GEA 1680 DKK
BLDPBD233814-1G BLDPBD233814-250MG BLDPBD233814-100MG
2-(Propan-2-yl)morpholine 95%
2-(Propan-2-yl)morpholine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₇H₁₅NO
Molekylvægt: 129,2 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD14582705
CAS-nummer: 89855-02-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR