4-Bromoindole-3-carboxyaldehyde 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD23329-1GEA 171 DKK
BLDPBD23329-1G BLDPBD23329-0.025G BLDPBD23329-10G BLDPBD23329-25G BLDPBD23329-250MG BLDPBD23329-5G
4-Bromoindole-3-carboxyaldehyde 98%
4-Bromoindole-3-carboxyaldehyde

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₉H₆BrNO
Molekylvægt: 224,06 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD05864695
CAS-nummer: 98600-34-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR