3-Amino-4-methoxybenzenesulphonic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD23324-500GEA 1010 DKK
BLDPBD23324-500G BLDPBD23324-100G BLDPBD23324-25G
3-Amino-4-methoxybenzenesulphonic acid 97%
3-Amino-4-methoxybenzenesulphonic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₉NO₄S
Molekylvægt: 203,2157 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00035759
CAS-nummer: 98-42-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR