4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD231138-1GEA 1370 DKK
BLDPBD231138-1G BLDPBD231138-250MG BLDPBD231138-100MG
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid 97%
4-((4,5-Dihydrothiazol-2-yl)carbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₁₁BN₂O₃S
Molekylvægt: 250,0800 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD06659881
CAS-nummer: 850568-30-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR