3-Bromo-N-methylbenzenesulphonamide 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD230536-1GEA
1480
DKK
BLDPBD230536-1G
BLDPBD230536-250MG
BLDPBD230536-100MG
3-Bromo-N-methylbenzenesulphonamide 98%
3-Bromo-N-methylbenzenesulphonamide
Storage condition: Sealed in dry, Room Temperature
Formel:
C₇H₈BrNO₂S Molekylvægt: 250,11 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07363818 CAS-nummer: 153435-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...