6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD230110-1GEA 1370 DKK
BLDPBD230110-1G BLDPBD230110-100MG BLDPBD230110-5G BLDPBD230110-250MG
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester 98%
6-(Boc-methylamino)-3-pyridineboronic acid pinacol ester

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₇H₂₇BN₂O₄
Molekylvægt: 334,22 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD09037484
CAS-nummer: 1032758-87-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR