1-Methylindole-3-boronic acid pinacol ester 96%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD227849-100MGEA 187 DKK
BLDPBD227849-100MG
1-Methylindole-3-boronic acid pinacol ester 96%
1-Methylindole-3-boronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₅H₂₀BNO₂
Molekylvægt: 257,1358 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD11894338
CAS-nummer: 683229-61-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR