2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD227574-1GEA 62.1 DKK
BLDPBD227574-1G BLDPBD227574-25G BLDPBD227574-10G BLDPBD227574-5G
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol 97%
2,6-Di(tert-butyl)-4-methylcyclohexan-1-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₃₀O
Molekylvægt: 226,398 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00278493
CAS-nummer: 163119-16-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR