6-Amino-1-naphthalenesulphonic acid 95%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD22701-100GEA 1070 DKK
BLDPBD22701-100G BLDPBD22701-25G BLDPBD22701-5G
6-Amino-1-naphthalenesulphonic acid 95%
6-Amino-1-naphthalenesulphonic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₀H₉NO₃S
Molekylvægt: 223,2500 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00035724
CAS-nummer: 81-05-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR