3,5-Dichloro-4-fluorobenzaldehyde 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD226661-1GEA
489
DKK
BLDPBD226661-1G
BLDPBD226661-25G
BLDPBD226661-250MG
BLDPBD226661-5G
3,5-Dichloro-4-fluorobenzaldehyde 98%
3,5-Dichloro-4-fluorobenzaldehyde
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₃Cl₂FO Molekylvægt: 193,0000 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11845864 CAS-nummer: 117820-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...