4-Carboxy-3-fluorophenylboronic acid pinacol ester 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD226552-1GEA
427
DKK
BLDPBD226552-1G
BLDPBD226552-5G
BLDPBD226552-250MG
4-Carboxy-3-fluorophenylboronic acid pinacol ester 98%
4-Carboxy-3-fluorophenylboronic acid pinacol ester
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₃H₁₆BFO₄ Molekylvægt: 266,0732 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09027284 CAS-nummer: 867256-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...