N-Boc-ethylenediamine hydrochloride 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD22640-25GEA
938
DKK
BLDPBD22640-25G
BLDPBD22640-250MG
BLDPBD22640-10G
BLDPBD22640-5G
BLDPBD22640-1G
N-Boc-ethylenediamine hydrochloride 95%
N-Boc-ethylenediamine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formel:
C₇H₁₇ClN₂O₂ Molekylvægt: 196,68 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD01076129 CAS-nummer: 79513-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...