4-Bromo-4'-iodobiphenyl 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD224606-100GEA 969 DKK
BLDPBD224606-100G BLDPBD224606-25G BLDPBD224606-10G BLDPBD224606-5G
4-Bromo-4'-iodobiphenyl 98%
4-Bromo-4'-iodobiphenyl

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₂H₈BrI
Molekylvægt: 359,00 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD17676204
CAS-nummer: 105946-82-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR