5-Amino-7-bromo-1H-indazole 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD224456-100MGEA 85.3 DKK
BLDPBD224456-100MG BLDPBD224456-250MG BLDPBD224456-1G BLDPBD224456-5G
5-Amino-7-bromo-1H-indazole 98%
5-Amino-7-bromo-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₆BrN₃
Molekylvægt: 212,0500 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08275114
CAS-nummer: 953411-10-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR