7-Hydroxychromone 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD223943-100MGEA 194 DKK
BLDPBD223943-100MG BLDPBD223943-1G BLDPBD223943-250MG
7-Hydroxychromone 97%
7-Hydroxychromone

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₆O₃
Molekylvægt: 162,1421 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00209371
CAS-nummer: 59887-89-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR