3-Bromophthalic anhydride 97%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD222548-25GEA 1950 DKK
BLDPBD222548-25G BLDPBD222548-5G BLDPBD222548-10G BLDPBD222548-1G BLDPBD222548-100G
3-Bromophthalic anhydride 97%
3-Bromophthalic anhydride

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₃BrO₃
Molekylvægt: 227,0116 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09800402
CAS-nummer: 82-73-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR