trans,trans-Dibenzylideneacetone 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD222314-10GEA 77.6 DKK
BLDPBD222314-10G BLDPBD222314-1G BLDPBD222314-100G BLDPBD222314-5G BLDPBD222314-500G BLDPBD222314-25G
trans,trans-Dibenzylideneacetone 98%
trans,trans-Dibenzylideneacetone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₁₇H₁₄O
Molekylvægt: 234,30 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004790
CAS-nummer: 35225-79-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR