Ethyl-6-chloropicolinate 95%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD220776-5GEA 287 DKK
BLDPBD220776-5G BLDPBD220776-25G BLDPBD220776-1G BLDPBD220776-100G
Ethyl-6-chloropicolinate 95%
Ethyl-6-chloropicolinate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₈H₈ClNO₂
Molekylvægt: 185,61 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09878438
CAS-nummer: 21190-89-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR