4-Amino-3-bromobenzoic acid 98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD21909-5GEA
225
DKK
BLDPBD21909-5G
BLDPBD21909-100G
BLDPBD21909-10G
BLDPBD21909-500G
BLDPBD21909-25G
BLDPBD21909-1G
4-Amino-3-bromobenzoic acid 98%
4-Amino-3-bromobenzoic acid
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formel:
C₇H₆BrNO₂ Molekylvægt: 216,0320 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03407439 CAS-nummer: 6311-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...