2-Methoxy-6-methylpyridine 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD21905-10GEA 132 DKK
BLDPBD21905-10G BLDPBD21905-25G BLDPBD21905-5G
2-Methoxy-6-methylpyridine 98%
2-Methoxy-6-methylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₉NO
Molekylvægt: 123,16 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD04112507
CAS-nummer: 63071-03-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR