Methyl 6-hydroxypyrimidine-4-carboxylate 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD218852-5GEA 1800 DKK
BLDPBD218852-5G BLDPBD218852-25G BLDPBD218852-1G BLDPBD218852-250MG
Methyl 6-hydroxypyrimidine-4-carboxylate 98%
Methyl 6-hydroxypyrimidine-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₆H₆N₂O₃
Molekylvægt: 154,123 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD21640297
CAS-nummer: 7399-93-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR