Methyl 3-oxo-2,3-dihydro-1H-isoindole-4-carboxylate 97%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD218560-100MGEA 628 DKK
BLDPBD218560-100MG
Methyl 3-oxo-2,3-dihydro-1H-isoindole-4-carboxylate 97%
Methyl 3-oxo-2,3-dihydro-1H-isoindole-4-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₀H₉NO₃
Molekylvægt: 191,183 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD11506113
CAS-nummer: 935269-25-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR