3-Carboxy-4-fluorophenylboronic acid pinacol ester 95%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
|
BLDPBD217976-5GEA
667
DKK
BLDPBD217976-5G
BLDPBD217976-25G
BLDPBD217976-1G
BLDPBD217976-100G
3-Carboxy-4-fluorophenylboronic acid pinacol ester 95%
3-Carboxy-4-fluorophenylboronic acid pinacol ester
Storage condition: Sealed in dry, Room Temperature
Formel:
C₁₃H₁₆BFO₄ Molekylvægt: 266,07 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD12546519 CAS-nummer: 882679-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...