2-Chloro-(5-ethoxycarbonyl)phenylboronic acid 98%

Leverandør: BLD PHARMATECH GMBH
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD217848-1GEA 876 DKK
BLDPBD217848-1G BLDPBD217848-5G BLDPBD217848-250MG
2-Chloro-(5-ethoxycarbonyl)phenylboronic acid 98%
2-Chloro-(5-ethoxycarbonyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Formel: C₉H₁₀BClO₄
Molekylvægt: 228,4373 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD08689529
CAS-nummer: 913835-93-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR