2-Ethylbenzoic acid 98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

BLDPBD21771-25GEA 365 DKK
BLDPBD21771-25G BLDPBD21771-100G BLDPBD21771-1G BLDPBD21771-5G
2-Ethylbenzoic acid 98%
2-Ethylbenzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₀O₂
Molekylvægt: 150,1700 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00045838
CAS-nummer: 612-19-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR